missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adiponectin Rat ELISA Kit

Product Code. 10682044
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10682044

Brand: Invitrogen™ KRP0041

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Sandwich ELISA Kit

The Rat Adiponectin ELISA quantitates Rat Adiponectin in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rat Adiponectin. Principle of the method The Rat Adiponectin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Adiponectin is a protein hormone that modulates a number of metabolic processes, including glucose regulation and fatty acid catabolism. Adiponectin is exclusively secreted from adipose tissue into the bloodstream and is very abundant in plasma relative to many hormones. The hormone plays a role in the suppression of the metabolic derangements that may result in type 2 diabetes, obesity, atherosclerosis and non-alcoholic fatty liver disease (NAFLD). Adiponectin is secreted into the bloodsteam where it accounts for approximately 0.01% of all plasma protein. Plasma concentrations reveal a sexual dimorphism, with females having higher levels than males. Levels of adiponectin are reduced in diabetics compared to non-diabetics. Weight reduction significantly increases circulating levels. Adiponectin automatically self-associates into larger structures. Adiponectin binds to a number of receptors. So far, two receptors have been identified, with homology to G protein-coupled receptors. Adiponectin exerts some of its weight reduction effects via the brain. This is similar to the action of leptin, but the two hormones perform complementary actions, and can have additive effects. Adiponectin affects glucose flux (gluconeogenesis, glucose uptake), lipid catabolism (beta-oxidation, triglyceride clearance), endothelial dysfunction, insulin sensitivity and weight loss. Administration of Adiponectin remains a promising area of research for clinical therapy in diseases such as obesity, type 2 diabetes and fatty liver disease.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_144744.3, NP_653345.1
Assay Range 0.375-24 ng/mL
Assay Sensitivity 50 pg/mL
Conjugate Biotin
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant
Target Species Rat
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 246253
Gene Symbol Adipoq
Incubation Time 3.3 hr.
Interassay CV 4.9%
Intraassay CV 4.8%
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene Adiponectin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma, 10 μL; Serum, 10 μL; Supernatant, 100 μL
Storage Requirements 2°C to 8°C
Target Rat
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 20 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.