missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ Zymogram, 10%, Protein Gel, 12-well
Click to view available options
Wells:
12-well
Unit Size:
Pack of 10
Description
Includes
Catalog numbers for gels refer to a single box. One box contains 10 gels.
- Proteases are run on a gel containing one of these substrates under denaturing conditions and visualized as clear bands against a dark background following a simple renaturing, developing, and staining protocol
- Zymogram gels are commonly used to detect matrix metalloproteinases
- Tris-Glycine gels with a substrate (gelatin or casein) incorporated into the gel
- Made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, highly purified water and casein or gelatin as the substrate; All have a 4% stacking gel
Sample Loading Volume: 20μL; 1D Well
- The 10% Zymogram (Gelatin) Gel is a 10% Tris-Glycine gel with 0.1% gelatin as the substrate
- Sensitivity Level: 10% Zymogram Gelatin Gel: 5 x 10-6 units of collagenase
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Gel Percentage | 10% |
| Gel Thickness | 1 mm |
| Gel Type | Zymogram |
| Product Type | Homogeneous Page |
| Separation of | Protein |
| Separation Range | 30 to 150 kDa |
| Wells | 12-well |
| Quantity | Each |
| Well Design | 1D Well |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction