Learn More
NOTUM Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14663-1-AP-150UL
This item is not returnable.
View return policy
Description
The NOTUM antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NOTUM. This antibody recognizes human, mouse antigen. The NOTUM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NOTUM | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 147111 | |
| IgG |
| Western Blot | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |