Learn More
NOTCH2/NOTCH2NL Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68212-1-IG-150UL
This item is not returnable.
View return policy
Description
The NOTCH2/NOTCH2NL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NOTCH2/NOTCH2NL. This antibody recognizes human antigen. The NOTCH2/NOTCH2NL antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| NOTCH2/NOTCH2NL | |
| Monoclonal | |
| 1000 μg/mL | |
| N2N | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 250-265 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2D5E1 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 388677 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |