missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOTCH2 (Cleaved Asp1733) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599443
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of fragment of activated Notch2 resulting from cleavage adjacent to Asp1733.
Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
Specifications
| NOTCH2 (Cleaved Asp1733) | |
| Polyclonal | |
| Unconjugated | |
| NOTCH2 | |
| AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18129, 29492, 4853 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35516, Q04721, Q9QW30 | |
| NOTCH2 | |
| A synthesized peptide derived from human NOTCH2(Accession Q04721), corresponding to amino acid residues L1717-S1741. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction