Learn More
GPE Scientific™ NorLoc™ NMR Tube Caps
Description
- Features a patented design that addresses many of the flaws in the traditional NMR tube cap.
- Can be applied much more easily and quickly, thereby increasing personal safety and saving valuable time.
- Incorporates an advanced seal design, conferring superior holding and sealing capabilities.
- Superior retention of the cap to the tube forms a vaulted seal that safeguards precious or critical samples, even during refrigerated cold storage and variable temperature cycling and withstands repeated removals and reapplications
Specifications
Specifications
| Product Type | NMR Tube Cap |
| Color | Blue Royal |
| Quantity | 100/Pk. |
| Size | 5 mm |