Learn More
Norfloxacin-d5 (ethyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Norfloxacin D5 |
| CAS | 1015856-57-1 |
| Molecular Formula | C16 2H5 H13 F N3 O3 |
| Quantity | 0.005 g |
| Synonym | 3-Quinolinecarboxylic acid, 1-(ethyl-1,1,2,2,2-d5)-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, 1-(Ethyl-1,1,2,2,2-d5)-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, Norfloxacin-d5, Norfloxacin D5 (ethyl D5) |
| InChI Formula | InChI=1 S/C16H18FN3O3/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20/h7-9,18 H,2-6H2,1H3,(H,22,23)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])N1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCNCC3 |
| IUPAC Name | 6-fluoro-4-oxo-1-(1,1,2,2,2-pentadeuterioethyl)-7-piperazin-1-ylquinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 324.36 |
| Formula Weight | 324.1646 g/mol |
| Show More |