Learn More
Invitrogen™ Norepinephrine Transporter Monoclonal Antibody (CL3063)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524647
Description
Immunogen sequence: SLYYLFSSFT LNLPWTDCGH TWNSPNCTDP KLLNGSVLGN HTKYSKYKFT PAAEFYERGV LHLHESSGIH DI Binds to an epitope located within the peptide sequence HTKYSKYKFTPAAEF as determined by overlapping synthetic peptides. Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.
Norepinephrine Transporter [NET, or noradrenaline transporter (NAT)] is a monoamine transporter that transports the neurotransmitter noradrenaline from the synapse back to its vesicles for storage until later use. It also appears to transport the neurotransmitter dopamine in the same way, but to a lesser degree. Studies have shown a decrease in NET levels in the locus coeruleus in patients diagnosed with major depression. Cocaine, amphetamines and many therapeutic antidepressants, such as the SNRIs (serotonin-norepinephrine reuptake inhibitors) and the tricyclic antidepressants (TCAs) act to raise noradrenaline. Furthermore, deficits in the NET gene have been associated with ADHD.
Specifications
| Norepinephrine Transporter | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O55192, P23975 | |
| SLC6A2 | |
| Recombinant protein fragment of Human SLC6A2 (Product #RP-102393) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| CL3063 | |
| Unconjugated | |
| SLC6A2 | |
| NaCl-dependent norepinephrine transporter; NAT1; NE transporter; NET; NE-T; NET1; neurotransmitter transporter; neurotransmitter transporter, noradrenal; NORADR; noradrenaline; noradrenaline transporter; norepinephrine transporter; Slc6a2; SLC6A5; sodium-dependent noradrenaline transporter; solute carrier family 6 (neurotransmitter transporter), member 2; solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2; solute carrier family 6 member 2; solute carrier family 6 member 5; transmembrane region 4..26 transmembrane region 55..77 transmembrane region 105..123 transmembrane region 136..157 | |
| Mouse | |
| Protein A | |
| RUO | |
| 20538, 6530, 83511 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.