Learn More
Invitrogen™ NOP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114297
This item is not returnable.
View return policy
Description
Immunogen sequence: PEQPFEKAAF QKQNDTPKGP QPPTVSPIRS SRPPPAKRKK SQSRGNSQLL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
NOP2 is a member of the unc-84 homolog family and encodes a nuclear nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Alternatively spliced transcript variants have been described.
Specifications
| NOP2 | |
| Polyclonal | |
| Unconjugated | |
| Nop2 | |
| 120kDa; 25S rRNA (cytosine(2870)-C(5))-methyltransferase; A530002O17; cb164; im:6893299; MGC117384; MGC149287; MGC149288; N2428; NOL1; NOL1/NOP2/Sun domain family, member 1; NOP120; NOP2; NOP2 nucleolar protein; NOP2 nucleolar protein homolog; NOP2 nucleolar protein homolog (yeast); nop2p; NSUN1; Nucleolar protein 1; nucleolar protein 1, 120 kDa; nucleolar protein 1, 120kDa; Nucleolar protein 2; Nucleolar protein 2 homolog; p120; probable 28S rRNA (cytosine(4447)-C(5))-methyltransferase; probable 28S rRNA (cytosine-C(5))-methyltransferase; proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2; rRNA (cytosine-C5-)-methyltransferase NOP2; sb:cb164; si:dkey-91i17.1; Unknown (protein for IMAGE:6909257); YNA1; YNL061W; YNL2428W | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4839 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P46087 | |
| Nop2 | |
| Synthetic peptide directed towards the C terminal region of human NOP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |