Learn More
Amarell™ Nonmercury Enclosed Scale Low Temperature Precision Thermometer
Click to view available options
Length (Metric):
350 mm
Unit Size:
Each
Description
- Enclosed milk glass scale
- Toluene or pentane compound filling with a clear or blue capillary back
- Richter-top finish with button
- In transparent plastic square case
- Suitable for government verification or calibration
Specifications
Specifications
| Certifications/Compliance | DIN 12 778 |
| Immersion Type | Total Immersion |
| Length (Metric) | 350 mm |
| Material | Glass |
| Thermometer Fill | Pentane Compound, Blue Capillary Back |
| Product Type | Low Temperature Thermometer |