Learn More
Amarell™ Nonmercury Enclosed Scale Low Temperature Precision Thermometer
Product Code. 10717981
Click to view available options
Length (Metric):
350 mm
Unit Size:
Each
Product Code. 10717981
Brand: Amarell™ L33018
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Precision thermometer for the measurement of extremely cold temperatures.
  • Enclosed milk glass scale
  • Toluene or pentane compound filling with a clear or blue capillary back
  • Richter-top finish with button
  • In transparent plastic square case
  • Suitable for government verification or calibration
Specifications
Certifications/Compliance DIN 12 778
Immersion Type Total Immersion
Length (Metric) 350 mm
Material Glass
Thermometer Fill Pentane Compound, Blue Capillary Back
Product Type Low Temperature Thermometer