missing translation for 'onlineSavingsMsg'
Learn More

Nomega-Amino-L-arginine hydrochloride, 97%, Thermo Scientific™

Product Code. p-7039567
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
This item is not returnable. View return policy

Product Code. 15475048

Brand: Thermo Scientific Alfa Aesar H58861.MF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1031799-40-2
Molecular Formula C6H16ClN5O2
Molecular Weight (g/mol) 225.68
MDL Number MFCD00672614
InChI Key YGVMJVSXFJRHGQ-UHFFFAOYNA-N
Synonym nomega-amino-l-arginine hydrochloride, 2s-2-amino-5-n'-aminocarbamimidamido pentanoic acid hydrochloride, ng-amino-l-arginine hydrochloride, c6h15n5o2.hcl, s-2-amino-5-hydrazinecarboximidamido pentanoic acid hydrochloride
PubChem CID 91886146
SMILES Cl.NNC(N)=NCCCC(N)C(O)=O

Specifications

IUPAC Name (2S)-2-amino-5-[[amino(hydrazinyl)methylidene]amino]pentanoic acid;hydrochloride
Quantity 50 mg
Formula Weight 225.67
Sensitivity Hygroscopic
Percent Purity 97%
Chemical Name or Material Nomega-Amino-L-arginine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.