missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3.1 Monoclonal Antibody (OTI4H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805190S
This item is not returnable.
View return policy
Description
NKX3.1 Monoclonal antibody specifically detects NKX3.1 in Human samples. It is validated for Western BlotSpecifications
| NKX3.1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10827 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7067 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI4H6 | |
| Unconjugated | |
| Thra | |
| bagpipe; BAPX2; Bax; Drosophila NK-3 transcription factor, locus 1; homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK3 homeobox 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NK-3 transcription factor, locus 1; NK-3 transcription factor, locus 1 (Drosophila); NKX3; NKX3.1; Nkx-3.1; Nkx3-1; NKX3-1 antibody; NKX3A; prostate-specific ag | |
| Human recombinant protein fragment corresponding to amino acids 1-220 of human THRA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction