Learn More
Nitric Acid, 4M, Chem Lab™
Product Code. 11973273
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11973273
Brand: Chem-Lab CL05.1912.1000
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Specifications
pH <1
Color Colorless
Concentration 252.04g HNO3/l H2O = 4 N
Density 1.07g/mL
Odor Odourless
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons compliance-icons
Product Identifier
  • Nitric acid 4 mol/l
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid