missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Nisin ReadyMade™ Solution

Product Code. 30044029
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044029 1 mL 1mL
30044053 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044029 Supplier TOKU-E™ Supplier No. N0841ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nisin ReadyMade™ Solution is provided as a sterile-filtered solution in 0.02 N HCl.

  • CAS Number: 1414-45-5
  • Molecular Formula: C143H230N42O37S7
  • Molecular Weight: 3,354.07 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Lactococcus lactis
  • Spectrum: Nisin is effective against Gram-positive bacteria, but has little or no effect on Gram-negative bacteria, yeasts, or fungi
  • Mechanism of Action: Nisin binds to lipid II which is a membrane-bound advanced intermediate involved in cell wall biosynthesis. It sequesters lipid II from its functional location. The nisin-lipid II complex leads to formation of pores in the bacterial membrane leading to bacterial cell death

Specifications

Species Bacteria
CAS 1414-45-5
Color Colorless
Common Name Nisin
Concentration 20,000 to 40,000 IU/mL
Content And Storage -20°C
Description Nisin ReadyMade™ Solution 20,000 to 40,000 IU/mL in 0.02 N HCl
Expression System Gram-positive bacteria
Formulation Liquid in 0.02 N HCl
For Use With (Application) Microbiology and Eukaryotic Cell Culture Applications
Molecular Weight (g/mol) 3354.07 g/mol
Product Type Antibiotic
Quantity 1 mL
Research Category Biomedicine, including cancer and veterinary research
Shipping Condition Ice Pack
Source Lactococcus lactis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.