Learn More
Nirogacestat, MedChemExpress
Product Code. 16476656
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476656 100 mg 100mg
16476348 10 mg 10mg
16468937 5 mg 5mg
16412467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16476656 Supplier Medchem Express Supplier No. HY15185
MedChemExpress Nirogacestat (PF-3084014) is a reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor with an IC50 of 6.2 nM. Inhibition of Notch signaling by Nirogacestat while minimizing gastrointestinal toxicity presents a promising approach for research of Notch receptor-dependent cancers.
Nirogacestat (PF-3084014) is a reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor with an IC50 of 6.2 nM. Inhibition of Notch signaling by Nirogacestat while minimizing gastrointestinal toxicity presents a promising approach for research of Notch receptor-dependent cancers.
Specifications
Chemical Name or Material Nirogacestat
CAS 1290543-63-3
Color White
Purity Grade Notes Research
Molecular Formula C27H41F2N5O
Quantity 100 mg
Synonym PF-3084014 PF-03084014
Solubility Information DMSO : 28.57 mg/mL (58.35 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES FC1=C2C(CC[C@H](N[C@@H](CCC)C(NC3=CN(C(C)(C)CNCC(C)(C)C)C=N3)=O)C2)=CC(F)=C1
Molecular Weight (g/mol) 489.64
Formula Weight 489.64
Percent Purity 97.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death