Learn More
Niraparib (R-enantiomer), MedChemExpress
Product Code. 16434078
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16434078
Brand: Medchem Express HY10619D
MedChemExpress Niraparib R-enantiomer (MK-4827 R-enantiomer) is an excellent PARP1 inhibitor with IC50 of 2.4 nM.
Niraparib R-enantiomer (MK-4827 R-enantiomer) is an excellent PARP1 inhibitor with IC50 of 2.4 nM.
Specifications
Chemical Name or Material Niraparib (R-enantiomer)
CAS 1038915-58-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H20N4O
Quantity 10 mg
Synonym MK 4827 (R-enantiomer)
Solubility Information DMSO : ≥ 32 mg/mL (99.88 mM)
SMILES O=C(C1=CC=CC2=CN(C3=CC=C([C@@H]4CNCCC4)C=C3)N=C12)N
Molecular Weight (g/mol) 320.39
Formula Weight 320.39
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death