missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acid, ≥99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
Specifications
Specifications
| Color | White |
| Assay Percent Range | ≥99% |
| Quantity | 100 g |
| Formula Weight | 123.1 |
| Vapor Pressure | 5.7 x 10-6mm Hg at 25°C (estimated)(Lit.) |
| Physical Form | Solid |
| Chemical Name or Material | Nicotinic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction