Learn More
Nickel plating solution, electroless, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
Description
For plating on silicon, germanium, gallium arsenide, cadmium sulfide, gold, nickel, iron and Kovar
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Nickel plating solution |
| Physical Form | Liquid |
| Color | Green |
| Quantity | 250 mL |
| Odor | Ammonia-like |
| MDL Number | MFCD00011137 |