Learn More
Nickel plating solution, electroless, ammonia free
Product Code. 11305158
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305158 250 mL 250mL
11395148 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11305158 Supplier Thermo Scientific Alfa Aesar Supplier No. 044070.AK
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nickel plating solution
Color Green
Name Note Electroless, ammonia free
Quantity 250 mL
MDL Number MFCD00011137
UN Number UN3082