Specifications
| Quantity | 5 Pk. |
| Product Type | Chattaway Spatula |
| Width (Metric) | 10 mm |
| Diameter (Metric) Outer | 3.6 mm |
| Dimensions (L x W) | 100 x 10 mm |
| Material | Nickel |
| Length (Metric) | 100 mm |