Learn More
Nicainoprol, MedChemExpress
Product Code. 16485817
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485817 5 mg 5mg
16400679 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16485817 Supplier Medchem Express Supplier No. HY100572
MedChemExpress Nicainoprol is a fast-sodium-channel blocking drug, which is a potent antiarrhythmic agent.
Nicainoprol is a fast-sodium-channel blocking drug, which is a potent antiarrhythmic agent.
Specifications
Chemical Name or Material Nicainoprol
CAS 76252-06-7
Color White
Purity Grade Notes Research
Molecular Formula C21H27N3O3
Quantity 5 mg
Synonym RU-42924
Solubility Information DMSO : ≥ 29 mg/mL (78.49 mM)
SMILES O=C(N1CCCC2=C1C(OCC(O)CNC(C)C)=CC=C2)C3=CC=CN=C3
Molecular Weight (g/mol) 369.46
Formula Weight 369.46
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation