Learn More
Tocris Bioscience™ NIBR 189
GREENER_CHOICE
Potent and selective EBI2 (GPR183) receptor antagonist
Brand:  Tocris Bioscience™ 5203/10
Encompass
Product Code. 15766576
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
NIBR 189 is a potent and selective EBI2 (GPR183) receptor antagonist (IC50 values are 11 and 15 nM for human and mouse EBI2 receptors respectively). Exhibits selectivity for EBI2 over a panel of other receptors, enzymes and transporters. Blocks oxysterol-induced migration of U937 monocyte cells. Orally bioavailable.
Specifications
Specifications
>99%
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions