Learn More
Tocris Bioscience™ NIBR 189
Potent and selective EBI2 (GPR183) receptor antagonist
Brand: Tocris Bioscience™ 5203/10
This item is not returnable.
View return policy
Description
NIBR 189 is a potent and selective EBI2 (GPR183) receptor antagonist (IC50 values are 11 and 15 nM for human and mouse EBI2 receptors respectively). Exhibits selectivity for EBI2 over a panel of other receptors, enzymes and transporters. Blocks oxysterol-induced migration of U937 monocyte cells. Orally bioavailable.Specifications
| >99% | |
| 10 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction