Learn More
Invitrogen™ NFKBIL1 Polyclonal Antibody

Description
Suggested positive control: HeLa. The amino acid sequence used as immunogen is 100% homologous in new world monkey, panda, mouse lemur, chimpanzee, orangutan, rhesus monkey, wild pig, 95% homologous in horse, rat, mouse, 89% homologous in frog, opossum, sea anemone and 74% homologous in platypus and carolina anole. Reacts with Frog. Immunogen sequence: RRYLRVQQVR WHPDRFLQR.
Specifications
Specifications
| Antigen | NFKBIL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | NFKBIL1 |
| Gene Accession No. | Q861W0, Q9UBC1 |
| Gene Alias | DADB-70P7.2; Def-7; ikappaBL; I-kappa-B-like protein; Ikbl; Inhibitor of kappa B-like protein; LST1; NF-kappa-B inhibitor-like protein 1; NFKB inhibitor like 1; NFKBIL; NFKBIL1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor like 1; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1 |
| Gene Symbols | NFKBIL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.