Learn More
NFIA Monoclonal Antibody (OTI6F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811216S
Encompass_Preferred
 View more versions of this product
Product Code. 17575034
  • 242.00 EUR / 30µL
Description
NFIA Monoclonal antibody specifically detects NFIA in Human samples. It is validated for Western Blot
Specifications
Specifications
NFIA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q12857
Mouse
Affinity chromatography
RUO
4774
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI6F1
Unconjugated
NFIA
1110047K16Rik; 9430022M17Rik; CCAAT-box-binding transcription factor; CTF; KIAA1439; NF1A; NF1-A; Nf1l21; NF-I/A; Nfia; NFI-A; NFI-L; Nuclear factor 1 A-type; nuclear factor 1/A; nuclear factor I A; nuclear factor I/A; TGGCA-binding protein; Unknown (protein for MGC:128467)
Human recombinant protein fragment corresponding to amino acids 200-509 of human NFIA produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions