Learn More
NFIA Monoclonal Antibody (OTI6F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811216S
This item is not returnable.
View return policy
Description
NFIA Monoclonal antibody specifically detects NFIA in Human samples. It is validated for Western BlotSpecifications
| NFIA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12857 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4774 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6F1 | |
| Unconjugated | |
| NFIA | |
| 1110047K16Rik; 9430022M17Rik; CCAAT-box-binding transcription factor; CTF; KIAA1439; NF1A; NF1-A; Nf1l21; NF-I/A; Nfia; NFI-A; NFI-L; Nuclear factor 1 A-type; nuclear factor 1/A; nuclear factor I A; nuclear factor I/A; TGGCA-binding protein; Unknown (protein for MGC:128467) | |
| Human recombinant protein fragment corresponding to amino acids 200-509 of human NFIA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |