missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-H Mouse anti-Human, Porcine, Clone: 1A3C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60331-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody recognize NEFH only.
Involved in the maintece of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).Specifications
| NF-H | |
| Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P12036, P12037 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100156492, 4744 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1A3C7 | |
| Unconjugated | |
| NEFH | |
| NEFH | |
| NF200 Fusion Protein Ag13517 | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction