Learn More
Eppendorf™ Test Tube Racks for Model C76 Water Bath Shaker
Click to view available options
Holds:
18.0 25 mm Tubes
29.0 16 mm Tubes
36.0 20 mm Tubes
Unit Size:
Each
Description
Require subplatform (14-284-46); order separately.
Specifications
Specifications
| For Use With (Equipment) | C76 water bath shaker |
| Holds | 36.0 20 mm Tubes |
| Includes | Insert |
| Material | Stainless Steel |