Learn More
Eppendorf™ Scientific Shaker Test Tube Platforms
Product Code. 10083144
Click to view available options
Tube Size:
12 to 15 mm
15 to 18 mm
18 to 21 mm
22 to 26 mm
26 to 30 mm
8 to 11 mm
No. of Positions:
20
22
30
42
48
60
Unit Size:
Each
Product Code. 10083144
Brand: Eppendorf™ M12890500
Only null left
Add to basket
Add to basket
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Tube Size 22 to 26 mm
Product Type Test Tube Rack
Material Stainless Steel
For Use With (Equipment) Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series
No. of Positions 22
Size Large