missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nevanimibe hydrochloride, MedChemExpress
Description
Nevanimibe hydrochloride (PD-132301 hydrochloride) is an orally active and selective acyl-coenzyme A:cholesterol O-acyltransferase 1 (ACAT1) inhibitor with an EC50 of 9 nM. Nevanimibe hydrochloride inhibits ACAT2 with an EC50 of 368 nM. Nevanimibe hydrochloride induces cell apoptosis and has the potential for adrenocortical cancer.
Specifications
Specifications
| Chemical Name or Material | Nevanimibe hydrochloride |
| CAS | 133825-81-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H40ClN3O |
| Quantity | 50 mg |
| Synonym | PD-132301 hydrochloride ATR101 hydrochloride |
| Solubility Information | DMSO : 41.67 mg/mL (90.97 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C(NCC1(C2=CC=C(N(C)C)C=C2)CCCC1)NC3=C(C(C)C)C=CC=C3C(C)C.[H]Cl |
| Molecular Weight (g/mol) | 458.08 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?