missing translation for 'onlineSavingsMsg'
Learn More

NeutraKine™ SCF Mouse anti-Human, Clone: 1B3C4, Proteintech

Product Code. 17659786 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17659786 25 μg 25µL
17649786 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17659786 Supplier Proteintech Supplier No. 690261IG25UG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NeutraKine™ SCF Monoclonal antibody apecifically detetects NeutraKine™ SCF antigen in Human Samples. It is validated for Neutralization, ELISA.

Spezifikation

Antigen NeutraKine™ SCF
Applications Neutralization, ELISA
Classification Monoclonal
Clone 1B3C4
Conjugate Unconjugated
Formulation Sterile PBS.
Gene Symbols KITLG
Host Species Mouse
Immunogen human Humankine SCF protein HZ-1024
Purification Method Protein G purification
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 4254
Target Species Human
Content And Storage Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Form Liquid
Isotype IgG1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.