Learn More
MP Biomedicals™ Neurofilament 68
Product Code. 11484166 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484166 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11484166 Supplier MP Biomedicals™ Supplier No. 08771092
Specifications
For Use With (Application) Cell Biology and Analysis
Molecular Weight (g/mol) 68
Name Neurofilament 68
Quantity 100 μg
Storage Requirements -20°C