missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuN Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26975-1-AP-150UL
This item is not returnable.
View return policy
Description
NeuN (Fox-3, NEUN, HRNBP3, RBFOX3, RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintece of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with maligt cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determit of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.Spezifikation
| NeuN | |
| Polyclonal | |
| Unconjugated | |
| RBFOX3 | |
| FOX3, Homo sapiens (Human), HRNBP3, NeuN, RBFOX3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110256156, 146713, 287847, 52897 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A6NFN3, Q8BIF2 | |
| Rbfox3 | |
| NeuN Fusion Protein Ag25689 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur