Learn More
Netupitant-d6, MedChemExpress
Description
Netupitant D6 is the deuterium labeled Netupitant (CID-6451149), which is a highly potent and selective, orally active neurokinin-1 (NK1) receptor antagonist.
Specifications
Specifications
| Chemical Name or Material | Netupitant-d6 |
| CAS | 2070015-31-3 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C30H26D6F6N4O |
| Color | White |
| Synonym | CID-6451149-d6 |
| SMILES | CN(CC1)CCN1C2=CC(C3=CC=CC=C3C)=C(N(C)C(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)=O)C=N2 |
| Industry Type | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 584.63 |
| Show More |