Learn More
Lovibond Tintometer™ Nessleriser Comparator
Click to view available options
Description:
Nessleriser 2150
Nessleriser 2250
Nessleriser 2250 Upgrade Kit
Includes:
Pair Nessler tubes, 250 mm with plungers
Stand
Stand, daylight lighting unit and pair Nessler tubes, 113 mm with plungers
Stand, daylight lighting unit and pair Nessler tubes, 250 mm with plungers
Unit Size:
Each
Description
Lovibond Tintometer™ Nessleriser Comparator is designed for color matching of pale-colored samples where a long path length is desirable for accurate results.
- Color comparator/colorimeter
- Usess comparator disc standards
- Determines chemical concentrations in solutions
- Available options:
- Tube length (250mm/113mm)
- Lighting (lighting unit or ambient light)
- DB 420 glass anti-meniscus plungers
Specifications
Specifications
| Description | Nessleriser 2250 |
| Includes | Stand |
| Type | Comparator |