Learn More
Bolle Safety Ness+ Platinum™ Safety Glasses
Ness+ Platinum™ Safety Glasses featuring all the benefits of the NESS model with added comfort from co-injected temples.
Brand:  Bolle Safety NESSPPSF
Product Code. 16436760
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Ultra-wraparound frame
  • PC/TPR co-injected temples for greater comfort
  • Panoramic field of vision
  • Scratch and fog resistant coating
  • Non-slip nose bridge
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions