Learn More
Neopentyl chloride, 98%
Product Code. 11367675
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367675 5 g 5g
11377675 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11367675 Supplier Thermo Scientific Alfa Aesar Supplier No. L02365.06
Request Bulk or Custom Format
CAS: 753-89-9 | C5H11Cl | 106.593 g/mol
Neopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Neopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and alkali metals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 753-89-9
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00000941
InChI Key JEKYMVBQWWZVHO-UHFFFAOYSA-N
PubChem CID 12956
IUPAC Name 1-chloro-2,2-dimethylpropane
SMILES CC(C)(C)CCl
Specifications
Melting Point -20°C
Density 0.868
Boiling Point 83°C to 84°C
Flash Point −8°C (17°F)
Odor Sweet
Linear Formula (CH3)3CCH2Cl
Refractive Index 1.4039
Quantity 5 g
UN Number UN1107
Beilstein 1730985
Synonym neopentyl chloride, propane, 1-chloro-2,2-dimethyl, neo-pentylchloride, t-c4h9ch2cl, 2,2-dimethylpropyl chloride, acmc-1bc1q, neopentyl chloride 5g, propane,1-chloro-2,2-dimethyl, 1-chloro-2,2-dimethylpropane
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 106.6
Percent Purity 98%
Chemical Name or Material Neopentyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Neopentyl chloride
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Neopentyl chloride