Learn More
Invitrogen™ Neon™ NxT Electroporation System 1-Channel Pipette
Description
The Neon NxT Electroporation System 1-Channel Pipette is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1S). Each Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S and NEON18SK) comes with a 1-channel pipette. This pipette is offered separately for those needing an additional 1-channel pipette or replacement for their existing pipette.
Specifications
Specifications
| Compatible Tips | Neon™ NxT 10-μL and 100-μL tips |
| Product Line | Neon NxT |
| Type | Electroporation Pipette |
| Volume (Metric) | 10, 100 μL |
| For Use With (Application) | Transfection |
| For Use With (Equipment) | Neon™ NxT Electroporation System |
| Quantity | 1 pipette |