Learn More
Dr.Weigert™ Neodisher LM 10
Description
- Alkaline detergent, liquid
- neodisher LM 10 is used for manual cleaning in plunge baths
- Mains ingredients : active chlorine and phosphates
- Suitable for precleaning items to be washed will then undergo a main mechanical cleaning process
- Remove radioactive contaminants and organic residues, even very stubborn
- Good material compatibility
- Dosage between 20-200 m/L
Specifications
Specifications
| Product Type | Neodisher LM |
| Form | Liquid |
| pH | 12.2 to 13.9 |
| Sterility | Non-sterile |
| Composition | Chlorine-based bleaching agents, phosphates |
| Concentration | 20 to 200 mL/L |
| Density | 1.2 g/cm3 |
| Description | 310 x 230 x 180 mm |
| Filtered | Filtered |
| Volume (Metric) | 10 L |
Safety and Handling
Product Identifier
- neodisher LM 10
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: potassium hydroxide,sodium hypochlorite