Learn More
Nemorubicin, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Nemorubicin (Methoxymorpholinyl doxorubicin) is a Doxorubicin derivative with potent antitumor activity. Nemorubicin is highly cytotoxic to a variety of tumor cell lines presenting a multidrug-resistant phenotype. Nemorubicin requirs an intact nucleotide excision repair (NER) system to exert its activity.
Specifications
Specifications
| Chemical Name or Material | Nemorubicin |
| CAS | 108852-90-0 |
| Color | Red |
| Purity Grade Notes | Research |
| Molecular Formula | C32H37NO13 |
| Quantity | 10 mg |
| Synonym | Methoxymorpholinyl doxorubicin FCE 23762 PNU 152243 |
| Solubility Information | DMSO : 65 mg/mL (100.99 mM; Need ultrasonic) |
| SMILES | COC1=C2C(C(C3=C(O)C(C[C@](C(CO)=O)(O)C[C@]4([H])O[C@H]5C[C@H](N6CCO[C@H](OC)C6)[C@H](O)[C@H](C)O5)=C4C(O)=C3C2=O)=O)=CC=C1 |
| Molecular Weight (g/mol) | 643.64 |
| Show More |