Learn More
Nemadectin, TRC
Specifications
Specifications
| Chemical Name or Material | Nemadectin |
| CAS | 102130-84-7 |
| Molecular Formula | C36 H52 O8 |
| Quantity | 25 mg |
| Synonym | Nemadectin, Milbemycin B','5-O-demethyl-28-deoxy-25-(1,3-dimethyl-1-butenyl)-6,28-epoxy-23-hydroxy-','[6R,23S,25S(E)]- (9CI), (6R,23S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-6,28-epoxy-23-hydroxymilbemycin B, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2 |
| InChI Formula | InChI=1S/C36H52O8/c1-20(2)13-24(6)32-25(7)30(37)18-35(44-32)17-28-16-27(43-35)12-11-22(4)14-21(3)9-8-10-26-19-41-33-31(38)23(5)15-29(34(39)42-28)36(26,33)40/h8-11,13,15,20-21,25,27-33,37-38,40H,12,14,16-19H2,1-7H3/b9-8+,22-11+,24-13+,26-10+/t21-,25-,27+,2 |
| SMILES | CC(C)\C=C(/C)\[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\C[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2)C[C@H](O)[C@@H]1C |
| Molecular Weight (g/mol) | 612.793 |
| Formula Weight | 612.366 |