Learn More
BRAND™ Needle-Valve Stopcock and Spare Spindle
Needle-Valve Stopcock and Spare Spindle constructed of PTFE with retention device.
Brand: BRAND™ 82170
17.05 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- Interchangeable
- PTFE valve spindle
- PP cap and retention device
- Valve opening 0 to 2.5 mm
Specifications
Valve Spindle | |
0 to 2.5 mm | |
PTFE | |
Retention Device |