missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD4 Antibody (S06-7C3), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 EUR - 645.00 EUR
Specifications
| Antigen | NEDD4 |
|---|---|
| Clone | S06-7C3 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648493
|
Novus Biologicals
NBP3-19815-100ul |
100 μg |
645.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657663
|
Novus Biologicals
NBP3-19815-50ul |
50 μg |
421.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NEDD4 Monoclonal antibody specifically detects NEDD4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NEDD4 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 | |
| A synthetic peptide of human NEDD4 (Uniprot # P46934) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-7C3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Neuroscience, Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 4734 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title