missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-4/PVRL4 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00 EUR - 349.00 EUR
Specifications
| Antigen | Nectin-4/PVRL4 |
|---|---|
| Clone | 2D4B6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236857
|
Proteintech
67721-1-IG-150UL |
150 μL |
410.00 EUR 349.00 EUR / 150µL Save 61.00 EUR 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233177
|
Proteintech
67721-1-IG-20UL |
20 μL |
145.00 EUR 129.00 EUR / 20µL Save 16.00 EUR 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Nectin-4/PVRL4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Nectin-4/PVRL4. This antibody recognizes human antigen. The Nectin-4/PVRL4 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Nectin-4/PVRL4 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 81607 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2D4B6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| LNIR; PRR4; nectin-4 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title