missing translation for 'onlineSavingsMsg'
Learn More

Necrostatin 2 S enantiomer, MedChemExpress

Product Code. 16448717
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448717 5 mg 5mg
16456118 10 mg 10mg
16442257 50 mg 50mg
16441429 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16448717 Supplier Medchem Express Supplier No. HY14622B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Necrostatin 2 S enantiomer is the S enantiomer of Necrostatin 2.

Necrostatin 2 S enantiomer is the S enantiomer of Necrostatin 2.

Specifications

Chemical Name or Material Necrostatin 2 S enantiomer
Physical Form Solid
CAS 852391-20-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H12ClN3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (180.04 mM; Need ultrasonic)
SMILES O=C1N(C)C([C@H](CC2=CNC3=C2C=CC=C3Cl)N1)=O
Molecular Weight (g/mol) 277.71
Formula Weight 277.71
Percent Purity 99.58%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.