missing translation for 'onlineSavingsMsg'
Learn More

NE 52-QQ57, MedChemExpress

Product Code. p-200097934
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479976

Brand: Medchem Express HY101784

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NE 52-QQ57 is a selective, and orally available GPR4 antagonist with an IC50 of 70 nM. NE 52-QQ57 has anti-inflammatory activity.

NE 52-QQ57 is a selective, and orally available GPR4 antagonist with an IC50 of 70 nM. NE 52-QQ57 has anti-inflammatory activity.

Specifications

Chemical Name or Material NE 52-QQ57
Physical Form Solid
CAS 1401728-56-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H28N6O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 20 mg/mL (48.02 mM; ultrasonic and warming and adjust pH to 5 with HCl and heat to 60°C)
SMILES CCC1=NN(C(C)=CC(C)=N2)C2=C1CC3=CC=C(C4=NN=C(C5CCNCC5)O4)C=C3
Molecular Weight (g/mol) 416.52
Formula Weight 416.52
Percent Purity 98.5%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.