missing translation for 'onlineSavingsMsg'
Learn More

NBTGR, MedChemExpress

Product Code. 16446897
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446897 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16446897 Supplier Medchem Express Supplier No. HY108322

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NBTGR (p-Nitrobenzylthioguanosine) is a potent inhibitor of nucleoside transport; inhibits adenosine uptake with a Ki of 70 nM.

NBTGR (p-Nitrobenzylthioguanosine) is a potent inhibitor of nucleoside transport

Specifications

Chemical Name or Material NBTGR
CAS 13153-27-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H18N6O6S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (230.19 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=C(N)N=C3SCC4=CC=C([N+]([O-])=O)C=C4)O1)O)O
Molecular Weight (g/mol) 434.43
Formula Weight 434.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.