Learn More
BRAND™ Natural Rubber Stoppers
Product Code. p-8010228 Shop All Brand Products
Click to view available options
Diameter (Metric) Top:
12 mm
14 mm
22 mm
55 mm
Unit Size:
Each
Pack of 25
Product Code. 15281439
Brand: BRAND™ 144384
Only null left
Add to basket
Add to basket
Protects liquids from outside contaminants. Provides a secure seal.
  • Natural rubber construction
  • Red color
  • Supplied in packs of 25
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 20 mm
Quantity 25/PK
Color Red
Diameter (Metric) Bottom 11 mm
Diameter (Metric) Top 14 mm
Material Natural Rubber