Learn More
Saint-Gobain Natural Rubber Stoppers
Click to view available options
Stopper Number:
38D
Unit Size:
Pack of 25
Description
- Temperature resistance from -35 to +70°C (120°C in humid conditions)
- DIN Standard 18271
- Gray-blue color
Specifications
Specifications
| Height (Metric) | 35mm |
| Quantity | 25/Pk. |
| Color | Blue |
| Diameter (Metric) Bottom | 31 mm |
| Diameter (Metric) Top | 38 mm |
| Material | Natural Rubber |
| Product Type | Rubber Stopper |
| Stopper Number | 38D |