missing translation for 'onlineSavingsMsg'
Learn More

Ark Plas Products™ Natural Kynar Standard Check Valves

Product Code. 11841170
Change view
Click to view available options
Description:
1/4 x 1/4 in.
1/8 x 1/8 in.
3/16 x 3/16 in.
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
11841170 1/4 x 1/4 in. Pack of 10
11831170 3/16 x 3/16 in. Pack of 10
11821170 1/8 x 1/8 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11841170 Supplier Ark Plas Products™ Supplier No. HAX24DF0
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these check valves for a variety of applications. Liebherr™ Natural Kynar Standard Check Valves feature a free floating design and can be used in any position with an immediate response to change in pressure or flow.

  • Molded in virgin materials only
  • Low cracking pressure
  • Disc based valves feature like bodies and can be sold in any configuration of barb, thread or luer

Specifications

Description 1/4 x 1/4 in.
Material Kynar PVDF
Min. Operating Temperature (Metric) 0°C
Packaging Quantity 10 Pack
Color Natural
Max. Operating Temperature (Metric) 80°C
Product Type Standard Check Valve
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.