Learn More
Reitenspiess™ Test Tube Brush with Wool Tip
Product Code. p-8001153
Click to view available options
Length (Metric):
220 mm
Unit Size:
Each
Product Code. 10031074
Brand: Reitenspiess™ 12220301
Only null left
Add to basket
Edge
Add to basket
Clean your test tubes using the Reitenspiess™ Test Tube Brush with Wool Tip. Use a test tube brush to clean your test tubes and other laboratory ware.
  • Natural bristles
  • Wool Tip
  • Sterile
Specifications
Length (Metric) 220 mm
Length (Metric) Bristle 80 mm
Length (Metric) Brush 105 mm
Quantity 1 Ea.