missing translation for 'onlineSavingsMsg'
Learn More

Scharlab NATRIUMTETRABORAT-10-HYDR.PA 500G

Brand:  Scharlab SO07070500


NATRIUMTETRABORAT-10-HYDR.PA 500G

Product Code. 11348574

  • ONLINE EXCLUSIVE PRICE
    194.25 EUR
    110.45 EUR / 500g
    Save 83.80 EUR
    43% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 500GR NATRIUMTETRABORAT-10-HYDR.PA

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : disodium tetraboratedecahydrate

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.